Learn More
Enzo Life Sciences Direct 8-iso-PGF2alpha ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900091
Encompass_Preferred
Highly sensititive ELISA, measuring 8-iso-Prostaglandin F.8-iso-PGF2α is produced in vivo by both non-cyclooxygenase and cyclooxygenase dependant mechanisms from arachidonic acid. It has been shown to be a potent vasoconstrictor, a potential mediator of hepatorenal syndrome and a mutagen in 3T3 cells and vascular smooth muscle cells. 8-iso-PGF2α has been shown to circulate in plasma and is excreted in urine. Alternative Name: 8-iso-Prostaglandin F2alpha. Sensitivity: 40.0 pg/ml (range 160 - 100,000 pg/ml). Assay Time: <3 hours or Overnight + 45 min. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR IgG Microtiter plate, Sample diluent, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Neutralizing reagent
Catalog No. 50-658-136