Learn More
Enzo Life Sciences 8-iso-PGF2alpha ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901010
Encompass_Preferred
The measurement of F2-isoprostanes has emerged as one of the most reliable approaches to assess oxidative stress status in vivo, providing an important tool to explore the role of oxidative stress in the pathogenesis of human disease. This is a colorimetric competitive immunoassay kit for the quantitative determination of free 8-iso-Prostaglandin F2α in biological fluids such as urine and in culture supernatants. To measure 8-iso-Prostaglandin F2α levels in serum, plasma or other tissue samples, use the Direct 8-iso-PGF2α EIA Kit. 8-iso-PGF2α is produced in vivo by both non-cyclooxygenase and cyclooxygenase dependant mechanisms from arachidonic acid. Alternative name: 8-iso-Prostaglandin F2alpha. Sensitivity: 16.3 pg/ml. Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Catalog No. 50-200-6884