Learn More
Enzo Life Sciences 8 iso PGF2 alpha polyclonal antibody (100 µl)
SDP
Catalog No. 50658245
Encompass_Preferred
Host: Rabbit. Immunogen: 8-iso-Prostaglandin F2α conjugated to porcine thyroglobulin. Source: Whole rabbit serum. Species Reactivity: Species independent. Applications: ELISA. Formulation: Liquid. Serum. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-658-245 Supplier Enzo Life Sciences Supplier No. ADI905015100
Host: Rabbit. Immunogen: 8-iso-Prostaglandin F2α conjugated to porcine thyroglobulin. Source: Whole rabbit serum. Species Reactivity: Species independent. Applications: ELISA. Formulation: Liquid. Serum. Handling: Avoid freeze/thaw cycles.