Learn More
Enzo Life Sciences 8-Isopentenylnaringenin (5mg). CAS: 68682-02-0
SDP
Supplier:  Enzo Life Sciences ALX385025M005
Encompass_Preferred
Prenyl flavonoid. Phytoestrogen. Selective, non-steroidal estrogen receptor α (ERα) ligand. Potent inhibitor of angiogenesis in vitro and in vivo. Chemopreventive agent against cancer induced by heterocyclic amines. Alternative name: 8-Prenylnaringenin. Purity: ≥98% (1H-NMR). Appearance: Light yellow powder. Solubility: Soluble in ethanol, methanol or DMSO. Nearly insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9053