Learn More
Cayman Chemical 8-Isoprostn AChE Tracer 100dtn
SDP
Catalog No. 502944159
Encompass_Preferred
A cardiac glycoside with cardiotonic and antiarrhythmic activities inhibits Na /K -ATPase in an isoform-specific biphasic manner with IC50 values of 0.298 and 410 UM for the high- and low affinity human erythrocyte isoenzymes respectively and 1.18 and 28.5 UM for the high- and low affinity porcine cerebral cortex isoenzymes respectively increases contractility and rhythmicity in isolated guinea pig heart in a concentration-dependent manner a degradation product of digoxin
Catalog No. 50-294-4159 Supplier Cayman Chemical Supplier No. 416350100 DTN
A cardiac glycoside with cardiotonic and antiarrhythmic activities inhibits Na /K -ATPase in an isoform-specific biphasic manner with IC50 values of 0.298 and 410 UM for the high- and low affinity human erythrocyte isoenzymes respectively and 1.18 and 28.5 UM for the high- and low affinity porcine cerebral cortex isoenzymes respectively increases contractility and rhythmicity in isolated guinea pig heart in a concentration-dependent manner a degradation product of digoxin