Learn More
Uline 8 Service Kit for Uline Poly Bag Sealer
Catalog No. NC0551219
Encompass
Maintenance and repair kit with Teflon coated 8 inch sealing bars, 3.5 inch cutter blade and pad, springs, pins, screws, and washers for H-163 tabletop impulse heat sealer allows periodic replacement of wear parts to maintain sealer performance.
Catalog No. NC0551219 Supplier Uline Supplier No. H164
Maintenance and repair kit with Teflon coated 8 inch sealing bars, 3.5 inch cutter blade and pad, springs, pins, screws, and washers for H-163 tabletop impulse heat sealer allows periodic replacement of wear parts to maintain sealer performance.