Learn More
Lcr Hallcrest 8 LEVEN THERMAX STRIPS RANGE B
Catalog No. NC9756310
Encompass_Preferred
This item has a minimum qty of 6 per supplier requirements.
8 Level Strips, Size: 51 by 18 mm (2 in. by 0.7 in. ), Type: Vertical, Self Adhesive, Scale: oC and oF, Range: Sold in units of 10, Degree C:71 - 110, degree F: 160 - 230
Catalog No. NC9756310 Supplier Lcr Hallcrest Supplier No. 08STHMRNGBD01PK
This item has a minimum qty of 6 per supplier requirements.
8 Level Strips, Size: 51 by 18 mm (2 in. by 0.7 in. ), Type: Vertical, Self Adhesive, Scale: oC and oF, Range: Sold in units of 10, Degree C:71 - 110, degree F: 160 - 230