Learn More
Medchemexpress LLC HY-101358 10mg Medchemexpress, 8-M-PDOT CAS:134865-70-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10135810MG
Encompass_Preferred
Medchemexpress, HY-101358 10mg 8-M-PDOT CAS:134865-70-6 8-M-PDOT (AH-002) is a selective melatonin MT2 receptor agonist. 8-M-PDOT is 5.2-fold selective for MT2 over MT1 receptors. 8-M-PDOT binds human recombinant MT2 and MT2 receptors with pKi values of 8.23 and 8.95 respectively. 8-M-PDOT has anxiolytic-like activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7836