Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 8-Mercaptooctanoic acid

Catalog No. 6750751G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-507-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-507-51G Supplier Sigma Aldrich Supplier No. 6750751G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

8-mercaptooctanoic acid (MOA) forms a self-assembled monolayer (SAM) on the surface of nanoparticles. MOA consists of thiol and carboxylic groups which helps in the surface treatment of different nanoparticles.This compound is used to produce hydrophilic SAMs. The resulting monolayers which are terminated with carboxylic acids can be further functionalized to introduce more complex end groups. Can also be used to form a protective barrier monolayer to allow deposition of a passivating oxide layer.This product should be stored in the freezer, 0 to -20°C

Specifications

CAS 74328-61-3
Density 1.0326 g/mL (at 25°C)
Molecular Formula C8H16O2S
Refractive Index n20/D 1.4808
Linear Formula C8H16O2S
Quantity 1 g
MDL Number MFCD08705279
Synonym 8-Thiooctanoic acid; MOA
Molecular Weight (g/mol) 176.28
Percent Purity 95%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.