Learn More
Quality Lab Accessories Qla 8 Mesh Basket Sinker, 316 Stainless Steel, Hinged End, Prevents Floating Dosage Forms, Each
Supplier:  Quality Lab Accessories BSK00801
Encompass
The QLA 8 Mesh Basket Sinker is a precision-engineered dissolution accessory made from 316 stainless steel, ideal for reliable drug release testing of larger capsules. Its innovative 8-mesh design prevents floating dosage forms, enhancing the accuracy and reproducibility of dissolution tests. Featuring a convenient hinged end for easy capsule introduction, this essential tool is compatible with various dissolution bath models from top manufacturers, ensuring versatility in laboratory settings.
Catalog No. NC9045328