Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Methoxypsoralen, MilliporeSigma™ Supelco™

Catalog No. 111012191 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2191 5 g
11-101-2190 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2191 Supplier MilliporeSigma Supelco Supplier No. M35015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 298-81-7
Physical Form Crystals
Linear Formula C12H8O4
MDL Number MFCD00005009
Melting Point 148°C to 150°C
Synonym 8-MOP; 9-Methoxyfuro[3,2-g][1 ]benzopyran-7-one; Ammoidin; Methoxsalen; Xanthotoxin
Solubility Soluble in acetic acid
Type Analytical Standard
Quantity 5 g
Formula Weight 216.19
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.