Learn More
Medchemexpress LLC 8-methyl-8-azabicyclo[3.2.1]octan-3-one | 532-24-1 | MFCD00005549 | 100.0% | 139.20 | C8H13NO | 10g
SDP
Catalog No. 5000268878
Encompass_Preferred
Tropinone is an organic synthesis intermediate and a skeleton for constructing derivatives containing thiazole rings and other structures Tropinone derivatives have anticancer activities such as inducing tumor cell apoptosis and inhibiting tyrosinase activity and can be used in research such as anticancer drug development and tyrosinase inhibitor design[1]
Catalog No. 50-002-68878 Supplier Medchemexpress LLC Supplier No. HYY013510G
Add to Cart
Add to Cart
Tropinone is an organic synthesis intermediate and a skeleton for constructing derivatives containing thiazole rings and other structures Tropinone derivatives have anticancer activities such as inducing tumor cell apoptosis and inhibiting tyrosinase activity and can be used in research such as anticancer drug development and tyrosinase inhibitor design[1]