Learn More
Medchemexpress LLC 8-methyl-8-azabicyclo[3.2.1]octan-3-yl 2-oxo-1,4-dihydroquinazoline-3-carboxylate hydrochloride | 131780-48-8 | MFCD00911718 | 99.0% | 351.83 g/mol | C17H22ClN3O3 | 5 MG
SDP
Catalog No. 5000242163
Encompass_Preferred
DAU 5884 hydrochloride is a potent, selective muscarinic M3 receptor antagonist provided as the hydrochloride salt for research use. It is used in vitro to study muscarinic receptor-mediated signaling, including methacholine-dependent cell proliferation and smooth muscle contractility.
  • Potent M3 receptor antagonist suitable for in vitro pharmacology.
  • Inhibits methacholine-dependent cell proliferation and muscle contractility.
  • High reported purity for reliable experimental results.
  • Available in small milligram quantities for screening and characterization.
  • Soluble in water and DMSO for diverse assay formats.
  • Provided as a hydrochloride salt for improved stability and handling.
Catalog No. 50-002-42163 Supplier Medchemexpress LLC Supplier No. HY1076455MG
DAU 5884 hydrochloride is a potent, selective muscarinic M3 receptor antagonist provided as the hydrochloride salt for research use. It is used in vitro to study muscarinic receptor-mediated signaling, including methacholine-dependent cell proliferation and smooth muscle contractility.
  • Potent M3 receptor antagonist suitable for in vitro pharmacology.
  • Inhibits methacholine-dependent cell proliferation and muscle contractility.
  • High reported purity for reliable experimental results.
  • Available in small milligram quantities for screening and characterization.
  • Soluble in water and DMSO for diverse assay formats.
  • Provided as a hydrochloride salt for improved stability and handling.