Learn More
Cayman Chemical 8-methylamino NAD sodium sa
SDP
Catalog No. 502917242
Encompass_Preferred
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations; has been used in the synthesis of cADPR derivatives
Catalog No. 50-291-7242 Supplier Cayman Chemical Supplier No. 38484700 UG
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations; has been used in the synthesis of cADPR derivatives