Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 8-Methylimidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

Catalog No. SYX001491G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX001491G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX001491G Supplier Sigma Aldrich Supplier No. SYX001491G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The reaction of 3,4-diethoxy-3-cyclobutene-1,2-dione (squaric acid diethyl ester) with glycosylamines (formed by reducing oligosaccharides) was studied
  • The derivatives formed were linked to amino-functionalized lipids, solids or proteins.3,4-dethoxy-3-cyclobutene-1,2-dione is used as a starting material for synthesis of furanones and quinones
  • It undergoes ethoxy substitution with amines and unsaturated organosilanes

Specifications

CAS 67625-40-5
Molecular Formula C11H12N2O2
Linear Formula C11H12N2O2
MDL Number MFCD03419470
Quantity 5 g
Molecular Weight (g/mol) 204.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.