Learn More
Covaris Inc 8 microTUBE-130 AFA Fiber H Slit Strip V2
SDP
Catalog No. NC1676980
Encompass
Application: FFPE Extraction, Chromatin Shearing, DNA/RNA Shearing. Sample Volume: 130 uL, and Fragment Size: 150 bp to 1.5 kb.
Catalog No. NC1676980 Supplier Covaris Inc Supplier No. 520239
Application: FFPE Extraction, Chromatin Shearing, DNA/RNA Shearing. Sample Volume: 130 uL, and Fragment Size: 150 bp to 1.5 kb.