Learn More
Covaris Inc 8 microTUBE-50 AFA Fiber H Slit Strip V2
SDP
Catalog No. NC1676979
Encompass
Specially designed vessel for use with Focused-ultrasonicators and small sample volumes. Fragment Size: 150 bp to 1.5 kb | Sample Volume: 50 μl | Application: DNA/RNA Shearing
Catalog No. NC1676979 Supplier Covaris Inc Supplier No. 520240
Specially designed vessel for use with Focused-ultrasonicators and small sample volumes. Fragment Size: 150 bp to 1.5 kb | Sample Volume: 50 μl | Application: DNA/RNA Shearing