Learn More
LGC GENOMICS LLC 8 ML COLUMN CHUCK
Catalog No. NC2310472
Encompass
Column holder for 8 mL syringe style synthesis columns (15 -30 µmol), MerMade 6 and 12 only. Use with column M8-1000, scale dependent on support loading.
Catalog No. NC2310472 Supplier LGC GENOMICS LLC Supplier No. AXM121024
Column holder for 8 mL syringe style synthesis columns (15 -30 µmol), MerMade 6 and 12 only. Use with column M8-1000, scale dependent on support loading.