Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-nitro-4-(3-nitrophenyl)-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline | 353495-22-4 | 99.4% | 5 MG
SDP

Catalog No. 5000233615
Encompass_Preferred

Phox-I2 is a small-molecule inhibitor that disrupts the p67phox-Rac1 interaction, inhibiting NADPH oxidase 2 (NOX2) activity and reducing reactive oxygen species (ROS) production. Supplied for research use, the compound is provided as a powder with defined purity and storage recommendations.

  • Cas: 353495-22-4
  • Chemical name: 8-nitro-4-(3-nitrophenyl)-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline
  • Chemical formula: C18H15N3O4
  • Molecular weight: 337.33 g/mol
  • Purity: 99.4%
  • Typical package size: 5 mg, other sizes available
  • Storage: powder at -20°C for long-term storage, 4°C for short-term storage
  • Intended use: research use only; not for human or clinical use

Catalog No. 50-002-33615 Supplier Medchemexpress LLC Supplier No. HY1195765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phox-I2 is a small-molecule inhibitor that disrupts the p67phox-Rac1 interaction, inhibiting NADPH oxidase 2 (NOX2) activity and reducing reactive oxygen species (ROS) production. Supplied for research use, the compound is provided as a powder with defined purity and storage recommendations.

  • Cas: 353495-22-4
  • Chemical name: 8-nitro-4-(3-nitrophenyl)-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline
  • Chemical formula: C18H15N3O4
  • Molecular weight: 337.33 g/mol
  • Purity: 99.4%
  • Typical package size: 5 mg, other sizes available
  • Storage: powder at -20°C for long-term storage, 4°C for short-term storage
  • Intended use: research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.