Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Nitroquinoline, 98%

Catalog No. p-7027092
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2357406 5 g
AAB2357414 25 g
AAB2357422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2357406 Supplier Thermo Scientific Chemicals Supplier No. B2357406
Only null left
Add to Cart

CAS: 607-35-2 | C9H6N2O2 | 174.159 g/mol

8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.

Solubility
Soluble in ethanol, ethyl ether and benzene, slightly soluble in cold water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-35-2
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00006806
InChI Key OQHHSGRZCKGLCY-UHFFFAOYSA-N
Synonym quinoline, 8-nitro, unii-ywo6s88v4v, ccris 457, ywo6s88v4v, 8-nitroqunioline, 4msn, 8-nitro-quinoline, gilstone-yellow, zlchem 249, pubchem5825
PubChem CID 11830
IUPAC Name 8-nitroquinoline
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])N=CC=C2

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
Beilstein 135178
Solubility Information Soluble in ethanol,ethyl ether and benzene,slightly soluble in cold water.
Formula Weight 174.16
Percent Purity 98%
Chemical Name or Material 8-Nitroquinoline
Hazard Category H315-H319-H335-H341-H351
Hazard Statement GHS H Statement
H341-H351-H315-H319-H335
Suspected of causing genetic defects.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 210-135-9
RTECSNumber VC1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.