Learn More
Medchemexpress LLC 8-Nonynoic acid | 30964-01-3 | 154.21 | 1 G
SDP
Catalog No. 5000401376
Encompass_Preferred
8-Nonynoic acid is a fatty acid that belongs to the group of octynoic acids. It exhibits antibacterial activity against gram-positive bacteria by binding to bacterial fatty acids, and inhibits the growth of gram-negative bacteria by inhibiting fatty acid synthesis. This compound is also capable of inhibiting the growth of both aerobic and anaerobic bacteria in biochemical assays.
  • Exhibits antibacterial activity against gram-positive bacteria.
  • Inhibits growth of gram-negative bacteria by inhibiting fatty acid synthesis.
  • Inhibits growth of both aerobic and anaerobic bacteria.
Catalog No. 50-004-01376 Supplier Medchemexpress LLC Supplier No. HY238481G
8-Nonynoic acid is a fatty acid that belongs to the group of octynoic acids. It exhibits antibacterial activity against gram-positive bacteria by binding to bacterial fatty acids, and inhibits the growth of gram-negative bacteria by inhibiting fatty acid synthesis. This compound is also capable of inhibiting the growth of both aerobic and anaerobic bacteria in biochemical assays.
  • Exhibits antibacterial activity against gram-positive bacteria.
  • Inhibits growth of gram-negative bacteria by inhibiting fatty acid synthesis.
  • Inhibits growth of both aerobic and anaerobic bacteria.