Learn More
Medchemexpress LLC 8-Nonynoic acid | 30964-01-3 | 154.21 | 250 MG
SDP
Catalog No. 5000402963
Encompass_Preferred
8-Nonynoic acid is a fatty acid that is part of the octynoic acids group. It demonstrates antibacterial properties against gram-positive bacteria by interacting with bacterial fatty acids. Additionally, it hinders the growth of gram-negative bacteria through the inhibition of fatty acid synthesis and can suppress the growth of both aerobic and anaerobic bacteria in biochemical assays.
  • Exhibits antibacterial activity against gram-positive bacteria.
  • Inhibits the growth of gram-negative bacteria by inhibiting fatty acid synthesis.
  • Capable of inhibiting the growth of both aerobic and anaerobic bacteria in biochemical assays.
Catalog No. 50-004-02963 Supplier Medchemexpress LLC Supplier No. 250MGHY23848
8-Nonynoic acid is a fatty acid that is part of the octynoic acids group. It demonstrates antibacterial properties against gram-positive bacteria by interacting with bacterial fatty acids. Additionally, it hinders the growth of gram-negative bacteria through the inhibition of fatty acid synthesis and can suppress the growth of both aerobic and anaerobic bacteria in biochemical assays.
  • Exhibits antibacterial activity against gram-positive bacteria.
  • Inhibits the growth of gram-negative bacteria by inhibiting fatty acid synthesis.
  • Capable of inhibiting the growth of both aerobic and anaerobic bacteria in biochemical assays.