Learn More
Medchemexpress LLC HY-N0757 5mg Medchemexpress, 8-​O-​Acetylharpagide CAS:6926-14-3 Purity:>98%
SDP
Catalog No. 502082818
Encompass_Preferred
Medchemexpress, HY-N0757 5mg 8-​O-​Acetylharpagide CAS:6926-14-3 8-O-Acetylharpagide is an iridoid isolated from Ajuga reptans with antitumoral, antiviral, antibacterial, and anti-inflammatory activities. 8-O-Acetylharpagide also has a biological activity on isolated smooth muscle preparations from guinea pig. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2818 Supplier Medchemexpress LLC Supplier No. HYN07575MG
Medchemexpress, HY-N0757 5mg 8-​O-​Acetylharpagide CAS:6926-14-3 8-O-Acetylharpagide is an iridoid isolated from Ajuga reptans with antitumoral, antiviral, antibacterial, and anti-inflammatory activities. 8-O-Acetylharpagide also has a biological activity on isolated smooth muscle preparations from guinea pig. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.