Learn More
AdipoGen 8-OH-6Me-pyrene-trisulfonamide
SDP
Supplier:  AdipoGen CDXH0043M050
Encompass_Preferred
Chemical. CAS 127044-59-1. Formula C22H25N3O7S3. MW 539.64. Synthetic. Fluorescent pH indicator for the weakly acidic pH range and enzyme substrate reference standard.
Catalog No. 50-256-0781