Learn More
Medchemexpress LLC 8-OH-DPAT 50.0MG
SDP
Catalog No. NC3806147
Encompass_Preferred
8-OH-DPAT is a potent and selective 5-HT agonist with a pIC50 of 8.19 for 5-HT1A and a Ki of 466 nM for 5-HT7 8-OH-DPAT weakly binds to 5-HT1B (pIC50 5.42) 5-HT (pIC50 less than 5).FOR RESEARCH ONLY.
Catalog No. NC3806147 Supplier Medchemexpress LLC Supplier No. HY112061
8-OH-DPAT is a potent and selective 5-HT agonist with a pIC50 of 8.19 for 5-HT1A and a Ki of 466 nM for 5-HT7 8-OH-DPAT weakly binds to 5-HT1B (pIC50 5.42) 5-HT (pIC50 less than 5).FOR RESEARCH ONLY.