Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-OH-DPAT | 78950-78-4 | 99.87% | 247.38 | 100 MG
SDP

Catalog No. 5000439368
Encompass_Preferred

8-OH-DPAT is a potent, brain-penetrant, and selective 5-HT agonist. It exhibits a pIC50 of 8.19 for 5-HT1A and a Ki of 466 nM for 5-HT7. It binds weakly to 5-HT1B (pIC50, 5.42) and 5-HT (pIC50 <5). For research use only.

  • Potent, brain-penetrant, and selective 5-HT agonist.
  • High affinity for 5-HT1A (pIC50 8.19) and 5-HT7 (Ki 466 nM).
  • Weakly binds to 5-HT1B (pIC50 5.42) and 5-HT (pIC50 <5). Does not bind to 5-HT6 or 5-HT4.
  • Normalizes hypolocomotion, increases wakefulness, and reduces REM sleep duration in orexin knockout (KO) mice, activating the 5-HT1A receptor.
  • Solid, white to light brown appearance.

Catalog No. 50-004-39368 Supplier Medchemexpress LLC Supplier No. HY112061100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-OH-DPAT is a potent, brain-penetrant, and selective 5-HT agonist. It exhibits a pIC50 of 8.19 for 5-HT1A and a Ki of 466 nM for 5-HT7. It binds weakly to 5-HT1B (pIC50, 5.42) and 5-HT (pIC50 <5). For research use only.

  • Potent, brain-penetrant, and selective 5-HT agonist.
  • High affinity for 5-HT1A (pIC50 8.19) and 5-HT7 (Ki 466 nM).
  • Weakly binds to 5-HT1B (pIC50 5.42) and 5-HT (pIC50 <5). Does not bind to 5-HT6 or 5-HT4.
  • Normalizes hypolocomotion, increases wakefulness, and reduces REM sleep duration in orexin knockout (KO) mice, activating the 5-HT1A receptor.
  • Solid, white to light brown appearance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.