Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 8-OHdG Competitive ELISA Kit

Catalog No. EEL004
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL004 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL004 Supplier Invitrogen™ Supplier No. EEL004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The multispecies 8-Hydroxydeoxyguanosine (8-OHdG) ELISA quantitates 8-OHdG in serum, plasma, and other biological fluids Principle of the method: The Competitive multispecies ELISA research-use-only kit is designed to quantitatively measure 8-Hydroxydeoxyguanosine (8-OHdG) independent of species. A 8-OHdG standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to 8-OHdG is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme-substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of 8-OHdG in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Oxygen is necessary for aerobic life, it can also participate in potentially toxic reactions involving oxygen free radicals and transition metals such as iron that damage membranes, proteins, and nucleic acids. n a normal human cell, there is a steady accumulation of DNA lesions with time. Substantial parts of these lesions are due to endogenous factors that damage DNA. These include reactive oxygens pecies derived from oxidative respiration. Additionally, malignant cells can produce hydrogen peroxide at levels as high as those characteristics for stimulated polymorphonuclear leukocytes. Some tumors may stimulate the defense systems of the body so that they react against the tumor to produce cytokines. Some cytokines can produce large amounts of ROS. 8-OHdG is one of the most widely studied lesions of this type. The presenceof 8-OHdG residues in DNA leads to GC to TA transversion unless repairs are made before DNA replication. Therefore, the presence of 8-OHdG may lead to mutagenesis. Furthermore, many observations indicate a direct correlation between 8-OHdG formation and carcinogenesis in vivo. It is generally accepted that the repair products of cellular DNA, for example 8-OHdG, are excreted into the urine without further metabolism. Therefore, 8-OHdG has been proposed as a urinary biomarker for in vivo oxidative DNA lesions.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.56-100 ng/mL
Assay Sensitivity 0.94 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Deoxyguanosine
Sample Type Plasma, Serum
Target Species All Species
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 8-Hydroxyguanosine; 8OHdDG8-Hydroxyguanosine; 8OHdDG;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target 8-OHdG
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.