Learn More
Cell Biolabs Inc OxiSelect™ Oxidative DNA Damage ELISA Kit (8-OHdG Quantitation), 96 assays
Supplier:  Cell Biolabs Inc STA320
Encompass_Preferred
Among numerous types of oxidative DNA damage, the formation of 8-hydroxydeoxyguanosine (8-OHdG) is a ubiquitous marker of oxidative stress. 8-OHdG, one of the oxidative DNA damage byproducts, is physiologically formed and enhanced by chemical carcinogens. During the repair of damaged DNA in vivo by exonucleases, the resulting 8-OH-dG is excreted without further metabolism into urine. The OxiSelect™ Oxidative DNA Damage ELISA Kit is a competitive enzyme immunoassay developed for rapid detection and quantitation of 8-OHdG in urine, serum, or other cell or tissue DNA samples. The quantity of 8-OHdG in unknown sample is determined by comparing its absorbance with that of a known 8-OHdG standard curve. The kit has an 8-OHdG detection sensitivity range of 100 pg/mL to 20 ng/mL. Each kit provides sufficient reagents to perform up to 96 assays, including standard curve and unknown samples.
Catalog No. NC9569610