Learn More
BioVendor, LLC 8-OHdG Check Human ELISA (Multispecies specificity), 96 wells
SDP
Supplier:  BioVendor, LLC N213100
Encompass
8-OHdG Check Human ELISA (Multispecies specificity), 96 wells. This 8OHdG Check immunoassay kit from Japan Institute For the Control of Aging (JaICA) is a competitive in vitro enzyme-linked immunosorbent assay for quantitative detection of the oxidative DNA adduct 8hydroxy2deoxyguanosine (8OHdG). For research use only. SAMPLE TYPE: urine, serum, plasma, tissue and other biological samples. SAMPLE REQUIREMENT: 50 l/well. CALIBRATION RANGE: 0.5200 ng/ml. SUMMARY: 8-OHdG is a widely used biomarker for the measurement of endogenous oxidative DNA damage. OTHER NAMES: 8-Hydroxy-2'-deoxyguanosine, 2'-Deoxy-7,8-dihydro-8-oxoguanosine.
Catalog No. 50-420-8