Learn More
BioVendor, LLC 8-OHdG Check (Ultrasensitive) Human ELISA (Multispecies specificity), 96 wells
SDP
Supplier:  BioVendor, LLC N213101
Encompass
8-OHdG Check (Ultrasensitive) Human ELISA (Multispecies specificity), 96 wells. The highly sensitive 8OHdG immunoassay kit from Japan Institute For the Control of Aging (JaICA) is a competitive in vitro enzyme-linked immunosorbent assay for quantitative measurement of the oxidative DNA adduct 8hydroxy2deoxyguanosine (8OHdG) in tissue, urine, serum, etc. For research use only. SAMPLE TYPE: tissue, urine, serum, etc.. SAMPLE REQUIREMENT: 50 l/well. CALIBRATION RANGE: 0.125 10 ng/ml. SUMMARY: 8-OHdG is a widely used biomarker for the measurement of endogenous oxidative DNA damage. OTHER NAMES: 8-Hydroxy-2'-deoxyguanosine, 2'-Deoxy-7,8-dihydro-8-oxoguanosine, high sensitivity
Catalog No. 50-420-9