Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%

Catalog No. AAH6382503
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6382503 1 g
AAH63825MD 250 mg
AAH6382506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6382503 Supplier Thermo Scientific Chemicals Supplier No. H6382503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 54745-74-3 | C6H12ClNO | 149.62 g/mol

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54745-74-3
Molecular Formula C6H12ClNO
Molecular Weight (g/mol) 149.62
MDL Number MFCD17926456,MFCD09800611
InChI Key XADOTNAXKKFKDY-UHFFFAOYNA-N
Synonym 8-oxa-3-azabicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclo 3,2,1 octane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane hcl, 8-oxa-3-azabicyclo 3.2.1 octanehydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane, hydrochloride, 8-oxa-3-aza-bicyclo 3.2.1 octane hydrochloride, 8-oxa-3-azabicyclooctane hydrochloride, 8-oxa-3-azabicyclo 3.2.1 octane-hcl, 8-oxa-3-aza-bicyclo 3.2.1 octane hcl
PubChem CID 21983536
SMILES Cl.C1CC2CNCC1O2

Specifications

Quantity 1 g
IUPAC Name 8-oxa-3-azabicyclo[3.2.1]octane;hydrochloride
Formula Weight 149.62
Percent Purity 97%
Chemical Name or Material 8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.