Learn More
Cusabio Technology LLC 8-oxoguanine DNA glycosylase
Catalog No. 5000051824
Encompass_Preferred
The Human 8-oxoguanine DNA glycosylase (hOGG1) ELISA Kit is designed for the quantitative detection of OGG1 in serum tissue homogenates and cell lysates This sandwich-based assay provides a detection range of 0 312-20 ng/mL with a sensitivity of 0 078 ng/mL ensuring accurate measurement of OGG1 levels in various biological samples The kit requires a sample volume of 50-100 L and delivers results within 1-5 hours using a 450 nm wavelength for absorbance measurement It is suitable for researchers studying DNA repair mechanisms oxidative stress and related pathologies offering reliable quantification of OGG1 to support experimental and diagnostic applications
Catalog No. 50-000-51824 Supplier Cusabio Technology LLC Supplier No. CSBE12686H
The Human 8-oxoguanine DNA glycosylase (hOGG1) ELISA Kit is designed for the quantitative detection of OGG1 in serum tissue homogenates and cell lysates This sandwich-based assay provides a detection range of 0 312-20 ng/mL with a sensitivity of 0 078 ng/mL ensuring accurate measurement of OGG1 levels in various biological samples The kit requires a sample volume of 50-100 L and delivers results within 1-5 hours using a 450 nm wavelength for absorbance measurement It is suitable for researchers studying DNA repair mechanisms oxidative stress and related pathologies offering reliable quantification of OGG1 to support experimental and diagnostic applications