Learn More
Electron Microscopy Sciences 8% Paraformaldehyde (formaldehyde) aqueous solution, 10x10 mL
SDP
Catalog No. 5025997
Encompass_Preferred
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 10 mL ampoules sealed under inert gas.
Catalog No. 50-259-97 Supplier Electron Microscopy Sciences Supplier No. 1578
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 10 mL ampoules sealed under inert gas.