Learn More
Enzo Life Sciences 8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphate . triethylammonium salt (5mg). CAS: 54364-02-2
SDP
Catalog No. 502009310
Encompass_Preferred
Selective activator of cGMP-dependent protein kinase Iα, Iβ and type II as well as of cGMP-gated ion channels. Higher activation potential as compared to cGMP; excellent cell membrane permeability and phosphodiesterase stability. Preferred alternative to unsuitable analogs such as dibutyryl-cGMP. Alternative name: 8-pCPT-cGMP . TEA. Purity: ≥99% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9310 Supplier Enzo Life Sciences Supplier No. ALX480031M005
Selective activator of cGMP-dependent protein kinase Iα, Iβ and type II as well as of cGMP-gated ion channels. Higher activation potential as compared to cGMP; excellent cell membrane permeability and phosphodiesterase stability. Preferred alternative to unsuitable analogs such as dibutyryl-cGMP. Alternative name: 8-pCPT-cGMP . TEA. Purity: ≥99% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water. Long Term Storage: -20°C.