Learn More
Enzo Life Sciences 8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphate . sodium salt (5mg). CAS: 51239-26-0
SDP
Catalog No. 502009321
Encompass_Preferred
Selective activator of cGMP-dependent protein kinase (PKG) Iα, Iβ, and type II as well as of cGMP-gated ion channels. Higher activation potential as compared to cGMP; excellent cell membrane permeability and phosphodiesterase stability. Preferred alternative to unsuitable analogs such as dibutyryl-cGMP. Alternative name: 8-pCPT-cGMP . Na. Purity: ≥99% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9321 Supplier Enzo Life Sciences Supplier No. ALX480088M005
Selective activator of cGMP-dependent protein kinase (PKG) Iα, Iβ, and type II as well as of cGMP-gated ion channels. Higher activation potential as compared to cGMP; excellent cell membrane permeability and phosphodiesterase stability. Preferred alternative to unsuitable analogs such as dibutyryl-cGMP. Alternative name: 8-pCPT-cGMP . Na. Purity: ≥99% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.