Learn More
Cayman Chemical 8-pCPT NAD sodIum salt 800ug
SDP
Catalog No. 502916927
Encompass_Preferred
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations
Catalog No. 50-291-6927 Supplier Cayman Chemical Supplier No. 38478800 UG
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations