Learn More
Sony Biotechnology Sony - 8 Peak Beads (5mL), 1 each
Catalog No. NC0883560
Encompass_Preferred
1 x l07 / mL of 3.0-3.4 um particles in deionized water with 0.02% Sodium Azide and 0.01% NP40
Catalog No. NC0883560 Supplier Sony Biotechnology Supplier No. AE700522
1 x l07 / mL of 3.0-3.4 um particles in deionized water with 0.02% Sodium Azide and 0.01% NP40