Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-phenyl-BODIPY 505/515 | 194235-40-0 | 324.18 g/mol | C19H19BF2N2 | 10 MG
SDP

Catalog No. 5000208942
Encompass_Preferred

8-Phenyl-BODIPY 505/515 is a phenyl-substituted BODIPY derivative fluorophore used as a fluorescent probe. It exhibits excitation around 502 nm and emission around 515 nm, and is supplied in small research quantities for labeling and imaging applications.

  • phenyl-substituted BODIPY fluorophore
  • excitation ~502 nm, emission ~515 nm
  • molecular weight 324.18 g/mol
  • chemical formula C19H19BF2N2
  • available in small vial sizes for research use

Catalog No. 50-002-08942 Supplier Medchemexpress LLC Supplier No. HYW08935310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Phenyl-BODIPY 505/515 is a phenyl-substituted BODIPY derivative fluorophore used as a fluorescent probe. It exhibits excitation around 502 nm and emission around 515 nm, and is supplied in small research quantities for labeling and imaging applications.

  • phenyl-substituted BODIPY fluorophore
  • excitation ~502 nm, emission ~515 nm
  • molecular weight 324.18 g/mol
  • chemical formula C19H19BF2N2
  • available in small vial sizes for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.