Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-phenyl-BODIPY 505/515 | 194235-40-0 | 98.5% | 324.18 | C19H19BF2N2 | 25 MG
SDP

Catalog No. 5000208769
Encompass_Preferred

8-Phenyl-BODIPY 505/515 is a phenyl-substituted boron-dipyrromethene (BODIPY) fluorophore supplied as a solid for use as a fluorescent probe. It absorbs in the green region and emits at ~515 nm, making it suitable for microscopy and flow cytometry. Molecular formula C19H19BF2N2; molecular weight 324.18 g·mol⁻1; CAS 194235-40-0.

  • Excitation 502 nm and emission 515 nm for green fluorescence applications.
  • High chemical purity, typically 98.5%, for reliable fluorescence performance.
  • Supplied as a solid in small quantities suitable for laboratory assays.
  • Suitable for fluorescence microscopy, flow cytometry, and other fluorescence-based assays.
  • Molecular formula C19H19BF2N2 and molecular weight 324.18 g·mol⁻1.

Catalog No. 50-002-08769 Supplier Medchemexpress LLC Supplier No. 25MGHYW089353
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Phenyl-BODIPY 505/515 is a phenyl-substituted boron-dipyrromethene (BODIPY) fluorophore supplied as a solid for use as a fluorescent probe. It absorbs in the green region and emits at ~515 nm, making it suitable for microscopy and flow cytometry. Molecular formula C19H19BF2N2; molecular weight 324.18 g·mol⁻1; CAS 194235-40-0.

  • Excitation 502 nm and emission 515 nm for green fluorescence applications.
  • High chemical purity, typically 98.5%, for reliable fluorescence performance.
  • Supplied as a solid in small quantities suitable for laboratory assays.
  • Suitable for fluorescence microscopy, flow cytometry, and other fluorescence-based assays.
  • Molecular formula C19H19BF2N2 and molecular weight 324.18 g·mol⁻1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.