Learn More
Cayman Chemical 8-phenylthio NAD sodium sal
SDP
Catalog No. 502917882
Encompass_Preferred
A derivative of NAD+; has been used in the determination of PARP target proteins
Catalog No. 50-291-7882 Supplier Cayman Chemical Supplier No. 38486800 UG
A derivative of NAD+; has been used in the determination of PARP target proteins