Learn More
FUJIFILM Biosciences 8 Phthalates Mixture
Catalog No. 5000450366
Encompass_Preferred
This product is a calibration reagent for the HM1000A Phthalate Ester Analyzer manufactured by Hitachi High-Tech Science Corporation Due to the manufacturing process some of the product may discolor to brown or black Please discard and use only the undiscolored portion It can also be used with the HM1000
Catalog No. 50-004-50366 Supplier FUJIFILM Biosciences Supplier No. 16828691
This product is a calibration reagent for the HM1000A Phthalate Ester Analyzer manufactured by Hitachi High-Tech Science Corporation Due to the manufacturing process some of the product may discolor to brown or black Please discard and use only the undiscolored portion It can also be used with the HM1000