Learn More
Electron Microscopy Sciences 8-Piece Stainless Steel Dissecting Kit
SDP
Catalog No. 5025545
Encompass_Preferred
This stainless steel 8 piece dissecting kit includes 1 4-1/2" dissecting forceps with curved and serrated tips, 1 curved dissecting needle, 1 glass pipette with rubber bulb, 1 4-1/2" dissecting forceps straight and serrated tips, 1 dissecting scalpel, 6" long, 1-1/2" cuting edge, 1 4-1/2" dissecting scissors, sharp, straight points, and 1 Plastic ruler 6", 1/16" div., millimeter and centimeter.
Catalog No. 50-255-45 Supplier Electron Microscopy Sciences Supplier No. 72950
This stainless steel 8 piece dissecting kit includes 1 4-1/2" dissecting forceps with curved and serrated tips, 1 curved dissecting needle, 1 glass pipette with rubber bulb, 1 4-1/2" dissecting forceps straight and serrated tips, 1 dissecting scalpel, 6" long, 1-1/2" cuting edge, 1 4-1/2" dissecting scissors, sharp, straight points, and 1 Plastic ruler 6", 1/16" div., millimeter and centimeter.