Learn More
Stellar Scientific 8 Place Magnetic Rack For 1.5mL Microcentrifuge Tubes
SDP
Catalog No. NC3933330
Encompass
Magnetic beads are used in a variety of lab processes including nucleic acid purification, cell-based assays, antibody purification, and protein purification.These high-quality anodized aluminum racks are excellent for microtubes, also called microcentrifuge tubes, and were design to optimize recovery of proteins or other biological structures.
Catalog No. NC3933330 Supplier Stellar Scientific Supplier No. 4EMR108
Magnetic beads are used in a variety of lab processes including nucleic acid purification, cell-based assays, antibody purification, and protein purification.These high-quality anodized aluminum racks are excellent for microtubes, also called microcentrifuge tubes, and were design to optimize recovery of proteins or other biological structures.