Learn More
Genesee Scientific Corporation 8-PORT MANIFOLDS
SDP
Catalog No. NC9980274
Encompass
CLP supplies an 8-port manifold which has a friction slip fit on to the 50ml combi-syringe. The tips on the manifold follow the standard 9mm center microplate spacing. With aqueous solutions this tip will provide equal dispensing across all 8 tips to within 1.7% of the dispensed volume. Because of accuracy concerns across larger diameters, CLP does not supply a twelve-port manifold. Made of 100% virgin polypropylene
Catalog No. NC9980274 Supplier Genesee Scientific Corporation Supplier No. 9621
Add to Cart
Add to Cart
CLP supplies an 8-port manifold which has a friction slip fit on to the 50ml combi-syringe. The tips on the manifold follow the standard 9mm center microplate spacing. With aqueous solutions this tip will provide equal dispensing across all 8 tips to within 1.7% of the dispensed volume. Because of accuracy concerns across larger diameters, CLP does not supply a twelve-port manifold. Made of 100% virgin polypropylene