Learn More
LUNA NANOTECH INC 8-Position Mini Magnetic Rack For 1.5 ML Tubes
Catalog No. 502566971
Encompass_Preferred
- Convenient Mini format magnetic racks for fast separation of magnetic beads in 1.5 mL microcentrifuge tubes. These racks can be used with our PuroMAG Mini kits as well as various magnetic beads offered by Luna Nanotech and other manufactureres.
- Angled design brings the tube bottom closer to the magnet to reduce magnetic separation time.
- Removable magnet strip allows resuspension of magnetic beads without the need to take the tubes out of the rack.
Catalog No. 50-256-6971 Supplier LUNA NANOTECH INC Supplier No. MGR008
- Convenient Mini format magnetic racks for fast separation of magnetic beads in 1.5 mL microcentrifuge tubes. These racks can be used with our PuroMAG Mini kits as well as various magnetic beads offered by Luna Nanotech and other manufactureres.
- Angled design brings the tube bottom closer to the magnet to reduce magnetic separation time.
- Removable magnet strip allows resuspension of magnetic beads without the need to take the tubes out of the rack.