Learn More
Organomation 8 Position S-EVAP-KD Evaporator
SDP
Catalog No. NC2301760
Encompass_Preferred
The digitally controlled 8 position S-EVAP-KD large scale solvent evaporator (Cat# 12018) can evaporate up to 8 samples at once with a capacity of up to 500 ml at each sample position.  Glassware sets can be purchased for individual solvent collection (Cat# GS2153), or central solvent collection (Cat# GS2153-C with Cat# GS2158-C8). Smaller glassware accommodating up to 250 ml can be purchased for individual solvent collection (Cat# GS2152), or central solvent collection (Cat# GS2152-C with Cat# GS2158-C8).  Smaller glassware is accommodated with the use adapter rings (Cat# XA2284).  
Catalog No. NC2301760 Supplier Organomation Supplier No. 12018
Add to Cart
Add to Cart
Request a Quote
The digitally controlled 8 position S-EVAP-KD large scale solvent evaporator (Cat# 12018) can evaporate up to 8 samples at once with a capacity of up to 500 ml at each sample position.  Glassware sets can be purchased for individual solvent collection (Cat# GS2153), or central solvent collection (Cat# GS2153-C with Cat# GS2158-C8). Smaller glassware accommodating up to 250 ml can be purchased for individual solvent collection (Cat# GS2152), or central solvent collection (Cat# GS2152-C with Cat# GS2158-C8).  Smaller glassware is accommodated with the use adapter rings (Cat# XA2284).  