Learn More
Analytical Sales & Services 8 Position Vial Tray for Covaris 8mL, 19x38mm (520089) Round Bottom Vials
Catalog No. 5000806771
Encompass
8 Position Vial Tray for Covaris 8mL, 19x38mm (520089) Round Bottom Vials\nAluminum vial tray with well bottoms designed to "hug" or conform to the shape of the vial to provide the most optimal temperature control (heating/cooling). Vials can be dropped in individually, or use a Covaris 500381 8-place vial holder rack to insert or remove 8 vials at a time. Can be used with automated platforms.\n\n• Holds (8) 8mL, 19x38mm Round Bottom Vials (Covaris 520089)\n• SLAS footprint\n• Corners tapped with 6-32 mounting holes for accessory attachment (i.e. TTD+)\n• Color: Purple\n\nQty/Pkg: Each\n\n* Corvaris vials and 8-place holder rack not included
Catalog No. 50-008-06771 Supplier Analytical Sales & Services Supplier No. 961935
8 Position Vial Tray for Covaris 8mL, 19x38mm (520089) Round Bottom Vials\nAluminum vial tray with well bottoms designed to "hug" or conform to the shape of the vial to provide the most optimal temperature control (heating/cooling). Vials can be dropped in individually, or use a Covaris 500381 8-place vial holder rack to insert or remove 8 vials at a time. Can be used with automated platforms.\n\n• Holds (8) 8mL, 19x38mm Round Bottom Vials (Covaris 520089)\n• SLAS footprint\n• Corners tapped with 6-32 mounting holes for accessory attachment (i.e. TTD+)\n• Color: Purple\n\nQty/Pkg: Each\n\n* Corvaris vials and 8-place holder rack not included