Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Prenylnaringenin | 53846-50-7 | 5 MG
SDP

Catalog No. 5000418378
Encompass_Preferred

8-Prenylnaringenin is an orally active prenyl flavonoid isolated from the hop spike Humulus lupulus. It activates the PI3K/Akt and AMPK pathways, upregulates OXPHOS complexes and Sirt1, and reduces ROS production and SOD activity. It improves muscle atrophy and obesity and inhibits angiogenesis. It also exhibits anticancer activity against glioblastoma and colon cancer, has LH/FSH regulatory activity, and may be used in bone health research.

  • Activates the PI3K/Akt pathway and the AMPK pathway
  • Upregulates OXPHOS complexes (II, III, and V) and Sirt1
  • Reduces ROS production and SOD activity
  • Improves muscle atrophy and obesity
  • Inhibits angiogenesis
  • Exhibits anticancer activity against glioblastoma and colon cancer
  • Has LH/FSH regulatory activity
  • May be used in bone health research
  • Purity: 99.63%
  • Appearance: solid, white to off-white

Catalog No. 50-004-18378 Supplier Medchemexpress LLC Supplier No. HYN27875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Prenylnaringenin is an orally active prenyl flavonoid isolated from the hop spike Humulus lupulus. It activates the PI3K/Akt and AMPK pathways, upregulates OXPHOS complexes and Sirt1, and reduces ROS production and SOD activity. It improves muscle atrophy and obesity and inhibits angiogenesis. It also exhibits anticancer activity against glioblastoma and colon cancer, has LH/FSH regulatory activity, and may be used in bone health research.

  • Activates the PI3K/Akt pathway and the AMPK pathway
  • Upregulates OXPHOS complexes (II, III, and V) and Sirt1
  • Reduces ROS production and SOD activity
  • Improves muscle atrophy and obesity
  • Inhibits angiogenesis
  • Exhibits anticancer activity against glioblastoma and colon cancer
  • Has LH/FSH regulatory activity
  • May be used in bone health research
  • Purity: 99.63%
  • Appearance: solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.