Learn More
Cayman Chemical 8-PrenylnarngenIn 10mg
SDP
Catalog No. 502979452
Encompass_Preferred
A prenylflavonoid with potent estrogenic activity that inhibits both ERα and ERβ (IC50s = 57 and 68 nM, respectively); effective in vivo, suppressing bone loss and temperature changes in rat models of osteoporosis and postmenopausal hot flashes
Catalog No. 50-297-9452 Supplier Cayman Chemical Supplier No. 1746210 MG
A prenylflavonoid with potent estrogenic activity that inhibits both ERα and ERβ (IC50s = 57 and 68 nM, respectively); effective in vivo, suppressing bone loss and temperature changes in rat models of osteoporosis and postmenopausal hot flashes