Learn More
Cayman Chemical 8-propylthio NAD sodium sal
SDP
Catalog No. 502917881
Encompass_Preferred
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations
Catalog No. 50-291-7881 Supplier Cayman Chemical Supplier No. 38488700 UG
A derivative of NAD+; has been used in the screening of analog-sensitive PARP1 gatekeeper mutations